Learn More
ATCC Rodent protoparvovirus 1; Strain: prototype (p)
Catalog No. 502382362
Encompass
Rodent protoparvovirus 1 strain Prototype (p) is propagated in A9 cells (ATCC CCL-1.4). This strain was plaque-purified from Minute virus of mice strain Crawford and deposited by Yale University School of Medicine.
Catalog No. 50-238-2362 Supplier ATCC Supplier No. VR1346
Rodent protoparvovirus 1 strain Prototype (p) is propagated in A9 cells (ATCC CCL-1.4). This strain was plaque-purified from Minute virus of mice strain Crawford and deposited by Yale University School of Medicine.