Learn More
Instech Rodent stand for ABS2 units with swivel lever arm and IV bag holder.
SDP
Catalog No. 5080985
Encompass
The Instech ABS2/R Stand is an optional accessory for the Instech ABS2™ automated blood sampler designed to position the sampler unit directly above a standard rodent cage. This configuration is intended to facilitate hand-free automated blood sampling from a swivel and tether system on freely-moving catherized rodents while minimizing tubing dead volume. The stand ensures proper positioning to reduce stress on the animal and minimize the length of the fluid lines, which helps with the accuracy of microsampling.
Features
    Stand: An X-shaped base with legs to elevate the sampler over a standard rodent cage.Counter-balanced Lever Arm: This arm attaches to the underside of the stand's intersection and holds the swivel, allowing the animal to move freely within the cage without tangling the tether.IV Bag Holder: A hook to hang an IV fluid bag (e.g., heparinized saline for KVO - Keep Vein Open flow).
Catalog No. 50-809-85 Supplier Instech Supplier No. ABS2/R
The Instech ABS2/R Stand is an optional accessory for the Instech ABS2™ automated blood sampler designed to position the sampler unit directly above a standard rodent cage. This configuration is intended to facilitate hand-free automated blood sampling from a swivel and tether system on freely-moving catherized rodents while minimizing tubing dead volume. The stand ensures proper positioning to reduce stress on the animal and minimize the length of the fluid lines, which helps with the accuracy of microsampling.
Features
    Stand: An X-shaped base with legs to elevate the sampler over a standard rodent cage.Counter-balanced Lever Arm: This arm attaches to the underside of the stand's intersection and holds the swivel, allowing the animal to move freely within the cage without tangling the tether.IV Bag Holder: A hook to hang an IV fluid bag (e.g., heparinized saline for KVO - Keep Vein Open flow).