Learn More
SAI Infusion Technologies Rodent Surgical Drapes with Press'N Seal Inserts
SDP
Catalog No. NC2381554
Encompass
15" x 19" Surgical Drape with an 11" x 12" Press'N Seal Insert. ETO Exposed for Sterility. Drapes are packaged 5/pk and are easy to remove one at a time.The Press'N Seal provides a transparent barrier for patient monitoring in rodent surgeries. Product retains body heat, hair, and allows for easy repositioning during surgery. For use as sterile surgical drape in preclinical studies. Product is for laboratory animal research only.
This item is not returnable. View return policy
Catalog No. NC2381554 Supplier SAI Infusion Technologies Supplier No. PSS51519
15" x 19" Surgical Drape with an 11" x 12" Press'N Seal Insert. ETO Exposed for Sterility. Drapes are packaged 5/pk and are easy to remove one at a time.The Press'N Seal provides a transparent barrier for patient monitoring in rodent surgeries. Product retains body heat, hair, and allows for easy repositioning during surgery. For use as sterile surgical drape in preclinical studies. Product is for laboratory animal research only.