Learn More
Braintree Scientific Rodent Workstand Heated Hard Pad Kit, requires pump adapter
SDP
Catalog No. 501955820
Encompass
Turn your workstand into a "The Heated Rodent WorkStand" with the new Heated Hard Pad kit. This Pad is designed to easily adapt to the Rodent Workstand. Its design will give you the ability to utilize the heated surface while in use on the work stand. The unit installs quite easily and can be ready for use in minutes with your existing or new Rodent WorkStand and circulating pump. The unit is also designed so it can be used as a stand-alone Heated Hard Pad. This product is a great versatile addition. (workstand not included) **Sold for research purposes only and not for human use**
Catalog No. 50-195-5820 Supplier Braintree Scientific Supplier No. RWHHP
Turn your workstand into a "The Heated Rodent WorkStand" with the new Heated Hard Pad kit. This Pad is designed to easily adapt to the Rodent Workstand. Its design will give you the ability to utilize the heated surface while in use on the work stand. The unit installs quite easily and can be ready for use in minutes with your existing or new Rodent WorkStand and circulating pump. The unit is also designed so it can be used as a stand-alone Heated Hard Pad. This product is a great versatile addition. (workstand not included) **Sold for research purposes only and not for human use**