Learn More
Grainger Rodenticide 4.1 lb 3 1/2 in H Green
Catalog No. 502767749
Encompass_Preferred
The TOMCAT Rodenticide Pail is a bait station containing rodenticide blocks to control mice and rats. It features a heavy-duty plastic pail with a child-resistant lid and locking bait compartment to keep bait secure. The pail contains eight 4oz all-weather paraffin bait blocks formulated with brodifacoum for effective rodent control. The blocks feature a peanut butter flavor that is highly palatable to rodents. The pail can be used indoors and outdoors and provides enough bait for a large area. The pail measures 14" diameter x 12" high.
Catalog No. 50-276-7749 Supplier Grainger Supplier No. 16W256
The TOMCAT Rodenticide Pail is a bait station containing rodenticide blocks to control mice and rats. It features a heavy-duty plastic pail with a child-resistant lid and locking bait compartment to keep bait secure. The pail contains eight 4oz all-weather paraffin bait blocks formulated with brodifacoum for effective rodent control. The blocks feature a peanut butter flavor that is highly palatable to rodents. The pail can be used indoors and outdoors and provides enough bait for a large area. The pail measures 14" diameter x 12" high.