Learn More
Medchemexpress LLC HY-17372 100mg , Rofecoxib CAS:162011-90-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY17372100MG
Encompass_Preferred
HY-17372 100mg Rofecoxib CAS: 162011-90-7 Rofecoxib is a potent, specific and orally active COX-2 inhibitor, with IC50s of 26 and 18 nM for human COX-2 in human osteosarcoma cells and Chinese hamster ovary cells, with a 1000-fold selectivity for COX-2 over human COX-1 (IC50 > 50 μM in U937 cells and > 15 μM in Chinese hamster ovary cells). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0204