Learn More
Cayman Chemical Roflumilast, Purity: ≥98%, CAS: 162401-32-3, 10 mg
SDP
Catalog No. 502971600
Encompass_Preferred
A PDE4 inhibitor (IC50 = 0.8 nM); selective for PDE4 over PDE1, -2, -3, and -5 (IC50s = >10, >10, >10, and 8 µM, respectively); inhibits fMLP-induced LTB4 production in isolated human neutrophils (IC50 = 2 nM); reduces the proliferation of anti-CD3 and anti-CD28-stimulated isolated human CD4+ T cells (IC30 = 7 nM); prevents pulmonary inflammatory cell infiltration and emphysema in mice exposed to cigarette smoke at 5 mg/kg
Catalog No. 50-297-1600 Supplier Cayman Chemical Supplier No. 1514110 MG
A PDE4 inhibitor (IC50 = 0.8 nM); selective for PDE4 over PDE1, -2, -3, and -5 (IC50s = >10, >10, >10, and 8 µM, respectively); inhibits fMLP-induced LTB4 production in isolated human neutrophils (IC50 = 2 nM); reduces the proliferation of anti-CD3 and anti-CD28-stimulated isolated human CD4+ T cells (IC30 = 7 nM); prevents pulmonary inflammatory cell infiltration and emphysema in mice exposed to cigarette smoke at 5 mg/kg