Learn More
Apexbio Technology LLC Roflumilast 162401-32-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062537
Encompass_Preferred
Roflumilast is an orally available selective inhibitor of phosphodiesterase-4 (PDE-4) with an IC50 value of approximately 0 11 nM PDE-4 enzymes catalyze the hydrolysis of cyclic adenosine monophosphate (cAMP) and are predominantly expressed in various inflammatory and immune cells By selectively inhibiting PDE-4 activity Roflumilast elevates intracellular cAMP levels thus regulating cellular inflammatory signaling pathways This mechanism supports its use in the reduction of inflammatory responses associated with chronic obstructive pulmonary disease (COPD) Additionally Roflumilast has been investigated experimentally regarding its potential influence on glucose metabolism in early-stage type 2 diabetes mellitus
Catalog No. 50-000-62537 Supplier Apexbio Technology LLC Supplier No. A43195.1
Roflumilast is an orally available selective inhibitor of phosphodiesterase-4 (PDE-4) with an IC50 value of approximately 0 11 nM PDE-4 enzymes catalyze the hydrolysis of cyclic adenosine monophosphate (cAMP) and are predominantly expressed in various inflammatory and immune cells By selectively inhibiting PDE-4 activity Roflumilast elevates intracellular cAMP levels thus regulating cellular inflammatory signaling pathways This mechanism supports its use in the reduction of inflammatory responses associated with chronic obstructive pulmonary disease (COPD) Additionally Roflumilast has been investigated experimentally regarding its potential influence on glucose metabolism in early-stage type 2 diabetes mellitus