Learn More
Medchemexpress LLC HY-15455 5mg Medchemexpress, Roflumilast CAS:162401-32-3 Purity:>98%
SDP
Catalog No. 502030629
Encompass_Preferred
Medchemexpress, HY-15455 5mg Roflumilast CAS:162401-32-3 Roflumilast is a selective PDE4 inhibitor with IC50s of 0.7, 0.9, 0.7, and 0.2 nM for PDE4A1, PDEA4, PDEB1, and PDEB2, respectively, without affecting PDE1, PDE2, PDE3 or PDE5 isoenzymes from various cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0629 Supplier Medchemexpress LLC Supplier No. HY154555MG
Medchemexpress, HY-15455 5mg Roflumilast CAS:162401-32-3 Roflumilast is a selective PDE4 inhibitor with IC50s of 0.7, 0.9, 0.7, and 0.2 nM for PDE4A1, PDEA4, PDEB1, and PDEB2, respectively, without affecting PDE1, PDE2, PDE3 or PDE5 isoenzymes from various cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.