Learn More
Medchemexpress LLC Roflumilast-d4 | 1398065-69-4 | C17H10D4Cl2F2N2O3 | 1 MG
SDP
Catalog No. 5000376447
Encompass_Preferred
Roflumilast-d4 is a deuterium-labeled selective PDE4 inhibitor, primarily used for research. It can be utilized as a tracer or an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS. Incorporating stable heavy isotopes into drug molecules, such as deuterium, aids in quantitation during drug development and can influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-76447 Supplier Medchemexpress LLC Supplier No. HY15455S1MG
Roflumilast-d4 is a deuterium-labeled selective PDE4 inhibitor, primarily used for research. It can be utilized as a tracer or an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS. Incorporating stable heavy isotopes into drug molecules, such as deuterium, aids in quantitation during drug development and can influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS