Learn More
Medchemexpress LLC Roflumilast-d4 N-Oxide | 1794760-31-8 | C17H10D4Cl2F2N2O4 | 100 UG
SDP
Catalog No. 5000376580
Encompass_Preferred
Roflumilast-d4 N-Oxide is a deuterium-labeled version of Roflumilast, a selective PDE4 inhibitor. This compound is valuable as a tracer and internal standard for quantitative analysis in drug development, particularly due to the influence of deuteration on pharmacokinetic and metabolic profiles.
  • Used as a tracer for quantitation
  • Can be an internal standard for analytical methods
  • Deuteration can influence pharmacokinetic and metabolic profiles
  • Selective PDE4 inhibitor
Catalog No. 50-003-76580 Supplier Medchemexpress LLC Supplier No. HY15455S1100UG
Roflumilast-d4 N-Oxide is a deuterium-labeled version of Roflumilast, a selective PDE4 inhibitor. This compound is valuable as a tracer and internal standard for quantitative analysis in drug development, particularly due to the influence of deuteration on pharmacokinetic and metabolic profiles.
  • Used as a tracer for quantitation
  • Can be an internal standard for analytical methods
  • Deuteration can influence pharmacokinetic and metabolic profiles
  • Selective PDE4 inhibitor