Learn More
Selleck Chemical LLC Roflumilast 5mg 162401-32-3 APTA 2217, B9302-107, BY 217, BYK 20869
SDP
Catalog No. 506647
Encompass_Preferred
Roflumilast (APTA 2217, B9302-107, BY 217, BYK 20869) is a selective inhibitor of PDE4 with IC50 of 0.2-4.3 nM in a cell-free assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-664-7 Supplier Selleck Chemical LLC Supplier No. S21315MG
Roflumilast (APTA 2217, B9302-107, BY 217, BYK 20869) is a selective inhibitor of PDE4 with IC50 of 0.2-4.3 nM in a cell-free assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.