Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Roflumilast N-oxide | 292135-78-5 | 99.5% | 419.21 g/mol | C17H14Cl2F2N2O4 | 1 ML
SDP

Supplier:  Medchemexpress LLC HY10063910MM 1ML

Encompass_Preferred

Roflumilast N-oxide is the N-oxide metabolite of roflumilast and functions as a phosphodiesterase 4 (PDE4) inhibitor. It is intended for research and analytical use and is provided in solution and solid forms suitable for biochemical assays and analytical standards.

  • Acts as a phosphodiesterase 4 inhibitor.
  • Supplied as a 10 mM solution in DMSO, 1 mL.
  • Available as solid samples in multiple milligram sizes.
  • Purity 99.5%.
  • CAS number 292135-78-5.
  • Molecular formula C17H14Cl2F2N2O4.
  • Molecular weight 419.21 g/mol.

Catalog No. 50-002-14280


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.