Learn More
Medchemexpress LLC Roflumilast N-oxide | 292135-78-5 | MFCD04112984 | 99.2% | 419.2 g·mol⁻¹ | C17H14Cl2F2N2O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY100639R50MG
Encompass_Preferred
Roflumilast N-oxide is the analytical reference standard of the active metabolite of roflumilast, a selective phosphodiesterase 4 (PDE4) inhibitor. It is provided as a characterized, high-purity solid for research and analytical applications including method development, calibration, and quantitative assays.
  • Analytical reference standard for method development and quantification.
  • High purity suitable for HPLC and LC-MS analysis.
  • Characterized chemical identity for reliable results.
  • Supplied as a weighed solid for accurate sample preparation.
  • Intended for research and analytical use only.
Catalog No. 50-002-13825