Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Roflupram 10mg | 1093412-18-0 | 10 MG
SDP

Catalog No. 5000229135
Encompass_Preferred

Roflupram is a selective, orally active, brain-penetrant phosphodiesterase 4 (PDE4) inhibitor used in preclinical research. It exhibits potent activity against human PDE4 catalytic domains and has been reported to reverse cognitive deficits and reduce production of pro-inflammatory factors.

  • Selective PDE4 inhibitory activity (IC50 26.2 nM).
  • High purity (99.45%).
  • Molecular formula C16H20F2O4; molecular weight 314.32 g·mol-1.
  • Soluble in DMSO (≈100 mg/mL) and compatible with several in vivo formulations.
  • Stable when stored under recommended conditions (pure form: -20°C for up to 3 years).

Catalog No. 50-002-29135 Supplier Medchemexpress LLC Supplier No. HY11538310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Roflupram is a selective, orally active, brain-penetrant phosphodiesterase 4 (PDE4) inhibitor used in preclinical research. It exhibits potent activity against human PDE4 catalytic domains and has been reported to reverse cognitive deficits and reduce production of pro-inflammatory factors.

  • Selective PDE4 inhibitory activity (IC50 26.2 nM).
  • High purity (99.45%).
  • Molecular formula C16H20F2O4; molecular weight 314.32 g·mol-1.
  • Soluble in DMSO (≈100 mg/mL) and compatible with several in vivo formulations.
  • Stable when stored under recommended conditions (pure form: -20°C for up to 3 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.