Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Roflupram 50mg | 1093412-18-0 | 50 MG
SDP

Catalog No. 5000307977
Encompass_Preferred

Roflupram is a selective, orally active, brain-penetrant phosphodiesterase 4 (PDE4) inhibitor used in preclinical research to study neuroinflammation, cognitive function, and PDE4-mediated signaling. It is supplied as a high-purity research chemical with documented solubility and storage recommendations suitable for in vitro and in vivo use.

  • Selective PDE4 inhibition with low nanomolar potency.
  • Brain-penetrant, suitable for CNS studies.
  • High purity appropriate for biochemical and cellular assays.
  • Soluble in DMSO; in vivo formulation protocols available.
  • Recommended storage conditions to maintain stability.

Catalog No. 50-003-07977 Supplier Medchemexpress LLC Supplier No. HY11538350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Roflupram is a selective, orally active, brain-penetrant phosphodiesterase 4 (PDE4) inhibitor used in preclinical research to study neuroinflammation, cognitive function, and PDE4-mediated signaling. It is supplied as a high-purity research chemical with documented solubility and storage recommendations suitable for in vitro and in vivo use.

  • Selective PDE4 inhibition with low nanomolar potency.
  • Brain-penetrant, suitable for CNS studies.
  • High purity appropriate for biochemical and cellular assays.
  • Soluble in DMSO; in vivo formulation protocols available.
  • Recommended storage conditions to maintain stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.