Learn More
Medchemexpress LLC Rogocekib (CTX-712) | 2144751-78-8 | 98.2% | C19H17FN8O2 | 100 MG
SDP
Catalog No. 5000353342
Encompass_Preferred
Rogocekib is an orally effective CLK 2 inhibitor with an IC50 of 1.4 nM, demonstrating anti-tumor activity. It is for research use only.
  • Strong inhibitory effect on the proliferation of human myeloid K562 and MV-4-11 cells
  • Exhibits anti-leukemia activity in primary acute myeloid leukemia (AML) cells
  • Inhibits the phosphorylation of various serine/arginine (SR) proteins
  • Reduces tumor size and increases survival rates in mouse models of myelodysplastic syndromes (MDS) and acute myeloid leukemia (AML)
Catalog No. 50-003-53342 Supplier Medchemexpress LLC Supplier No. HY144875100MG
Rogocekib is an orally effective CLK 2 inhibitor with an IC50 of 1.4 nM, demonstrating anti-tumor activity. It is for research use only.
  • Strong inhibitory effect on the proliferation of human myeloid K562 and MV-4-11 cells
  • Exhibits anti-leukemia activity in primary acute myeloid leukemia (AML) cells
  • Inhibits the phosphorylation of various serine/arginine (SR) proteins
  • Reduces tumor size and increases survival rates in mouse models of myelodysplastic syndromes (MDS) and acute myeloid leukemia (AML)