Learn More
Eurofins Discoverx ROKβ/ROCK-I, active, 250 µg. (14-601M)
Supplier:  Eurofins Discoverx 14601M
Encompass
N-terminal 6xHis-tagged recombinant, human ROCK-I amino acids 17-535; auto activated by incubating with Mg/ATP and redialysed to remove excess ATP. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9032