Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rolapitant (hydrochloride hydrate) | 914462-92-3 | 98.4% | 554.95 | 25 MG
SDP

Catalog No. 5000392788
Encompass_Preferred

Rolapitant (hydrochloride hydrate) | 914462-92-3 | 98.4% | 554.95 | 25 MG

Catalog No. 50-003-92788 Supplier Medchemexpress LLC Supplier No. 25MGHY16436
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rolapitant (hydrochloride hydrate) | 914462-92-3 | 98.4% | 554.95 | 25 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.