Learn More
Medchemexpress LLC HY-16900 100mg , Rolipram (R,S)-Rolipram;SB 95952;ZK 62711 CAS:61413-54-5 Purity:98%
SDP
Catalog No. 501492805
Encompass_Preferred
Medchemexpress, HY-16900 100mg Rolipram (R,S)-Rolipram;SB 95952;ZK 62711 CAS:61413-54-5 Formula:C16H21NO3 IC50: 3 nM (PDE4A), 130 nM (PDE4B), 240 nM (PDE4D) Purity:98% Rolipram is a selective inhibitor of phosphodiesterases PDE4 with IC 50 of 3 nM, 130 nM and 240 nM for PDE4A, PDE4B, and PDE4D, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2805 Supplier Medchemexpress LLC Supplier No. HY16900100MG
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-16900 100mg Rolipram (R,S)-Rolipram;SB 95952;ZK 62711 CAS:61413-54-5 Formula:C16H21NO3 IC50: 3 nM (PDE4A), 130 nM (PDE4B), 240 nM (PDE4D) Purity:98% Rolipram is a selective inhibitor of phosphodiesterases PDE4 with IC 50 of 3 nM, 130 nM and 240 nM for PDE4A, PDE4B, and PDE4D, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.