Learn More
Enzo Life Sciences Rolipram (50mg). CAS: 61413-54-5
SDP
Supplier:  Enzo Life Sciences BMLPD1750050
Encompass_Preferred
Selective, cell permeable inhibitor of cAMP-specific phosphodiesterase (PDE IV, IC50=1 µM). A rolipram insensitive PDE IV subtype has been described. Alternative name: (+/-)-4-[3-(Cyclopentyloxy)-4-methoxyphenyl]-2-pyrrolidinone. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml, warm), 100% ethanol (25mg/ml, warm) or acetone. Long Term Storage: Ambient.
Catalog No. 50-201-1388