Learn More
Medchemexpress LLC HY-16900 50mg , Rolipram CAS:61413-54-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1690050MG
Encompass_Preferred
HY-16900 50mg Rolipram CAS: 61413-54-5 Rolipram is a selective phosphodiesterases PDE4 inhibitor with IC50s of 3 nM, 130 nM and 240 nM for PDE4A, PDE4B, and PDE4D, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0196