Learn More
Medchemexpress LLC Rolitetracycline | 751-97-3 | ≥98.0% | 527.57 | 1 ML
SDP
Catalog No. 5000387671
Encompass_Preferred
Rolitetracycline is a highly soluble, broad-spectrum antibiotic derived from tetracycline. It inhibits HIV-1 integrase, blocks Aβ fibril formation, and suppresses dengue virus proliferation. It mediates the inhibition of Aβ fibrils via a specific three-dimensional pharmacophore conformation and exhibits bacteriostatic and bactericidal activities. It acts synergistically with Penicillin G or Cephalothin and is a tool compound for studying bacterial infections (urinary tract infections, sepsis), HIV-1 and dengue virus infections, and Alzheimer's disease.
  • Highly soluble, broad-spectrum antibiotic
  • Inhibits HIV-1 integrase
  • Blocks Aβ fibril formation
  • Suppresses dengue virus proliferation
  • Bacteriostatic and bactericidal
  • Acts synergistically with Penicillin G or Cephalothin
  • Tool compound for studying bacterial infections, HIV-1, dengue virus infections, and Alzheimer's disease
Catalog No. 50-003-87671 Supplier Medchemexpress LLC Supplier No. HY1825710MM/1ML
Rolitetracycline is a highly soluble, broad-spectrum antibiotic derived from tetracycline. It inhibits HIV-1 integrase, blocks Aβ fibril formation, and suppresses dengue virus proliferation. It mediates the inhibition of Aβ fibrils via a specific three-dimensional pharmacophore conformation and exhibits bacteriostatic and bactericidal activities. It acts synergistically with Penicillin G or Cephalothin and is a tool compound for studying bacterial infections (urinary tract infections, sepsis), HIV-1 and dengue virus infections, and Alzheimer's disease.
  • Highly soluble, broad-spectrum antibiotic
  • Inhibits HIV-1 integrase
  • Blocks Aβ fibril formation
  • Suppresses dengue virus proliferation
  • Bacteriostatic and bactericidal
  • Acts synergistically with Penicillin G or Cephalothin
  • Tool compound for studying bacterial infections, HIV-1, dengue virus infections, and Alzheimer's disease