Learn More
Med Vet International Dynarex Stretch Gauze Bandage Roll, 3", 96/Case
Catalog No. 502831043
Encompass_Preferred
Dynarex Stretch Gauze Bandages are comfortable bandages ideal for primary and secondary wound care in both professional and home environments. Unique knit construction conforms to the body and offers ideal compression strength without constricting the area. Unique knit construction offers proper compression without constriction Highly absorbent to keep the wound site clean Requires minimal taping and conforms to difficult body contours Non-linting and ravel-free Ideal for both primary and secondary wound care dressings 4.1 yards, stretched
Catalog No. 50-283-1043 Supplier Med Vet International Supplier No. 3103
Dynarex Stretch Gauze Bandages are comfortable bandages ideal for primary and secondary wound care in both professional and home environments. Unique knit construction conforms to the body and offers ideal compression strength without constricting the area. Unique knit construction offers proper compression without constriction Highly absorbent to keep the wound site clean Requires minimal taping and conforms to difficult body contours Non-linting and ravel-free Ideal for both primary and secondary wound care dressings 4.1 yards, stretched