Learn More
Med Vet International Dynarex Stretch Gauze Bandage Roll, Sterile, 4", 96/Case
Catalog No. 502830871
Encompass_Preferred
Sterile gauze bandages ideal for both primary and secondary wound care dressings. Unique knit construction offers proper compression without constriction Self adhering; stays in place with minimal taping Conforms to difficult body contours Highly absorbent gauze roll is non-linting and ravel free to keep the wound site clean 4.1 yards, stretched Ideal for both primary and secondary wound care dressings Packed in peel-open poly pouches for aseptic handling
Catalog No. 50-283-0871 Supplier Med Vet International Supplier No. 3114
Sterile gauze bandages ideal for both primary and secondary wound care dressings. Unique knit construction offers proper compression without constriction Self adhering; stays in place with minimal taping Conforms to difficult body contours Highly absorbent gauze roll is non-linting and ravel free to keep the wound site clean 4.1 yards, stretched Ideal for both primary and secondary wound care dressings Packed in peel-open poly pouches for aseptic handling