Learn More
Brady™ Roll Form Pipe Marker: COLD WATER
Catalog No. 19112285 Shop All Brady Corporation Products
Change view
Click to view available options
Display Text:
COLD WATER
Catalog No. Display Text
19-112-285 COLD WATER
1 options
Catalog No. 19-112-285 Supplier Brady™ Supplier No. 41512
  • $137.00
    $107.65 / Each of 1
    Save $29.35
    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Provide clear notice to workers by identifying pipe contents and flow without the hassle of precise measurement around a pipe's diameter. Brady™ Roll Form Pipe Markers are made of Pressure Sensitive Vinyl and display text with clear color contrast.
  • Made of self-sticking vinyl suitable for indoor or outdoor applications
  • Legend repeats 65 times per roll
Specifications
Adhesive Type Acrylic, Permanent, Cold-temperature
Color White, Green
Format Roll
Material Pressure-sensitive Vinyl (B-946)
Product Type Roll Form Pipe Marker
Display Text COLD WATER
Size (English) 30 ft. x 8 in.
Thickness (English) 0.004 in.