Learn More
Sigma Aldrich Fine Chemicals Biosciences Roll SealTM sealing films1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z6901631EA
Encompass
Excel Scientific now offers adhesive sealing films on Roll-Seal rolls for use with high-throughput automated microplate sealers. film rolls constructed on three-inch-diameter plastic cores are compatible with most common adhesive sealers (Brandel Thermo-fisher/Abgene fluidX KBiosystems). The Roll-Seal format provides reliable and efficient sealing at a lower cost per plate with minimal user intervention. Robust static-free packaging provides protection during shipment and facilitates accurate alignment and adhesion.fast efficient high-throughput sealing 2.5-5 seconds/plate no handling and backing removal from individual sheetsHands-free reduces chance of mishap or contamination more uniform sealing from plate-to-plateHeat-free no high temperatures to damage sensitive samples or warp plates and cause robotic handling problemsEspecially convenient when working with foils avoids curling that occurs with individual sheetsEconomical lower cost per plate than
Catalog No. 50-290-5673