Learn More
Alkali Scientific Adhesive Roll-Stock™ for Automated Microplate Sealer, SilverSeal, 80mm x 100 meters (3.15" x 328’), 3" Cores
SDP
Catalog No. 502389112
Encompass_Preferred
Adhesive Roll-Stock™ for Automated Microplate and foils are ideal for use with high-throughput automated microplate sealers. The Roll-Stock format provides reliable and efficient sealing at a lower cost per plate with minimal user intervention. Includes: 80 mm wide x 100 meters long; seals approximately 830 microplates. Easy-to-pierce for direct sample recovery. Application: PCR, long-term storage, light sensitive assays & robotics. Temperature Range: -80°C to +120°C.
Catalog No. 50-238-9112 Supplier Alkali Scientific Supplier No. RS4SS
Adhesive Roll-Stock™ for Automated Microplate and foils are ideal for use with high-throughput automated microplate sealers. The Roll-Stock format provides reliable and efficient sealing at a lower cost per plate with minimal user intervention. Includes: 80 mm wide x 100 meters long; seals approximately 830 microplates. Easy-to-pierce for direct sample recovery. Application: PCR, long-term storage, light sensitive assays & robotics. Temperature Range: -80°C to +120°C.