Learn More
Alkali Scientific Adhesive Roll-Stock™ for Automated Microplate Sealer, ThermaSeal™, 79mm x 65 meters (3.093" x 214’), 3" Cores
SDP
Catalog No. 502389248
Encompass_Preferred
Adhesive Roll-Stock™ for Automated Microplate and foils are ideal for use with high-throughput automated microplate sealers. The Roll-Stock format provides reliable and efficient sealing at a lower cost per plate with minimal user intervention. Includes: 79 mm wide x 65 meters long; seals approximately 540 microplates. Features a pressure-activated, non-tacky, silicone adhesive. Application: qPCR, long-term storage, protein crystallography & HPLC. Temperature Range: -80°C to +105°C.
Catalog No. 50-238-9248 Supplier Alkali Scientific Supplier No. RS4TS
Adhesive Roll-Stock™ for Automated Microplate and foils are ideal for use with high-throughput automated microplate sealers. The Roll-Stock format provides reliable and efficient sealing at a lower cost per plate with minimal user intervention. Includes: 79 mm wide x 65 meters long; seals approximately 540 microplates. Features a pressure-activated, non-tacky, silicone adhesive. Application: qPCR, long-term storage, protein crystallography & HPLC. Temperature Range: -80°C to +105°C.