Learn More
Alkali Scientific Adhesive Roll-Stock™ for Automated Microplate Sealer, VaporSeal™, 81mm x 100 meters (3.2" x 328’), 3" Cores
SDP
Catalog No. 502389250
Encompass_Preferred
Adhesive Roll-Stock™ for Automated Microplate and foils are ideal for use with high-throughput automated microplate sealers. The Roll-Stock format provides reliable and efficient sealing at a lower cost per plate with minimal user intervention. Includes: 81 mm wide x 100 meters long; seals approximately 830 microplates. Designed to facilitate air and CO2 exchange. Application: Cell, tissue, and bacterial culture. Temperature Range: -20°C to +80°C.
Catalog No. 50-238-9250 Supplier Alkali Scientific Supplier No. RS4VP
Adhesive Roll-Stock™ for Automated Microplate and foils are ideal for use with high-throughput automated microplate sealers. The Roll-Stock format provides reliable and efficient sealing at a lower cost per plate with minimal user intervention. Includes: 81 mm wide x 100 meters long; seals approximately 830 microplates. Designed to facilitate air and CO2 exchange. Application: Cell, tissue, and bacterial culture. Temperature Range: -20°C to +80°C.