Learn More
Beckman Coulter ROLLER 4IN WIDE SOFTRUBBER
Catalog No. NC9517619
Encompass
Soft Rubber Roller 4 in available for easy lid placement and to ensure complete sealing
Catalog No. NC9517619 Supplier Beckman Coulter Supplier No. 538618
Soft Rubber Roller 4 in available for easy lid placement and to ensure complete sealing