Learn More
Bellco Glass, Inc. Digital Bottom RA 8Dk,230V- BB With Battery Backup
SDP
Catalog No. 501945860
Encompass_Preferred
The Bellco Digital Bottom Drive Roller Apparatus holds up to 50 (see specifications below) full size glass or plastic roller bottles (110 to 120 mm diameter and up to 510 mm in length).Digital bottom drive roller apparatus. Digital display speed indicator readout. 24v brushless dc motor and controller. Locking speed control knob. Optional battery backup gives up to 18 hours of battery power during powerInterruptions. Battery level indicator. Rotational switch for cw & ccw rotation.Optional motion alarm (factory installation only). MADE IN THE USA
Catalog No. 50-194-5860 Supplier Bellco Glass, Inc. Supplier No. 738008230
Add to Cart
Add to Cart
Request a Quote
The Bellco Digital Bottom Drive Roller Apparatus holds up to 50 (see specifications below) full size glass or plastic roller bottles (110 to 120 mm diameter and up to 510 mm in length).Digital bottom drive roller apparatus. Digital display speed indicator readout. 24v brushless dc motor and controller. Locking speed control knob. Optional battery backup gives up to 18 hours of battery power during powerInterruptions. Battery level indicator. Rotational switch for cw & ccw rotation.Optional motion alarm (factory installation only). MADE IN THE USA