Learn More
Bellco Glass, Inc. Digital Bottom Roller Apparatus 9Dk,230V W/O Battery Backup
SDP
Catalog No. 501945847
Encompass_Preferred
The Bellco Process Controlled Bottom Drive Roller Apparatus holds up to 50 (see specifications below) full size glass or plastic roller bottles (110 to 120 mm diameter and up to 510 mm in length). The unit features a brushless DC motor and controller that operate at bottle speeds ranging from 0.3 to 9.0 rpm with the ability to select clockwise or counter clockwise direction. Units feature brushless, maintenance-free DC motors with drive assemblies designed for quiet operation. Four corner bumpers and two locking castors (on the front wheel positions) are also provided. Rollers are easily removed for replacement by using the special tool provided with each unit. MADE IN THE USAAll-Position Drive facilitates the use of lightweight plastic roller bottles by ensuring rotation at each bottle station, independent of any load configurations.
Catalog No. 50-194-5847 Supplier Bellco Glass, Inc. Supplier No. 734009230
Add to Cart
Add to Cart
Request a Quote
The Bellco Process Controlled Bottom Drive Roller Apparatus holds up to 50 (see specifications below) full size glass or plastic roller bottles (110 to 120 mm diameter and up to 510 mm in length). The unit features a brushless DC motor and controller that operate at bottle speeds ranging from 0.3 to 9.0 rpm with the ability to select clockwise or counter clockwise direction. Units feature brushless, maintenance-free DC motors with drive assemblies designed for quiet operation. Four corner bumpers and two locking castors (on the front wheel positions) are also provided. Rollers are easily removed for replacement by using the special tool provided with each unit. MADE IN THE USAAll-Position Drive facilitates the use of lightweight plastic roller bottles by ensuring rotation at each bottle station, independent of any load configurations.