Learn More
Bellco Glass, Inc. Digital Top Dr 86P,115V W/O Battery Backup
SDP
Catalog No. 501945878
Encompass_Preferred
The Bellco Top Drive Roller Apparatus has been designed for production of animal cells, viruses and cell products. The unit accommodates 53 bottle positions with over 85,000 cm2 of surface area. Rollers have a key design to eliminate slipping and are easily removable for servicing of bearings. Rollers are formulated from a chemical-resistant non-slip EDPM. All units incorporate a solid state control with a brushless motor and controller, providing smooth operation and easy maintenance. All units feature rubber bumpers, handles, a slotted rear guard, and All Position Drive. The All Position Drive is designed to accommodate plastic roller bottles by preventing the slipping of the light-weight bottles. An optional Motion Alarm System provides audible and visual alarms should the primary fail. MADE IN THE USA
Catalog No. 50-194-5878 Supplier Bellco Glass, Inc. Supplier No. 746010115
Add to Cart
Add to Cart
Request a Quote
The Bellco Top Drive Roller Apparatus has been designed for production of animal cells, viruses and cell products. The unit accommodates 53 bottle positions with over 85,000 cm2 of surface area. Rollers have a key design to eliminate slipping and are easily removable for servicing of bearings. Rollers are formulated from a chemical-resistant non-slip EDPM. All units incorporate a solid state control with a brushless motor and controller, providing smooth operation and easy maintenance. All units feature rubber bumpers, handles, a slotted rear guard, and All Position Drive. The All Position Drive is designed to accommodate plastic roller bottles by preventing the slipping of the light-weight bottles. An optional Motion Alarm System provides audible and visual alarms should the primary fail. MADE IN THE USA