Learn More
Bellco Glass, Inc. Process Top Dr 86P,115V-W/BB With Battery Backup
SDP
Supplier:  Bellco Glass, Inc. 742020115
Encompass_Preferred
The Bellco Process Controlled Top Drive Roller Apparatus has a capacity of either 53 or 86 full size glass or plastic roller bottles (110 to 120 mm diameter and up to 510 mm in length), depending upon model. The unit features a brushless DC motor and controller that operate at bottle speeds ranging from 0.3 to 9.0 rpm with the ability to select clockwise or counter clockwise direction. Four corner bumpers and two locking castors (on the front wheel positions) are also provided. Rollers are easily removed for replacement by using the special tool provided with each unit. All-Position Drive facilitates the use of lightweight plastic roller bottles by ensuring rotation at each bottle station, independent of any load configurations. A fused power inlet module makes fuse replacement easy and also permits the user to completely disconnect the power cord when the unit is cleaned or relocated. MADE IN THE USA
Catalog No. 50-194-5870
Add to Cart
Request a Quote