Learn More
MVE BIOLOGICAL SOLUTIONS MVE Roller Base, 5-Wheel Dolly, 20.0 in I.D. (Internal Diameter)
Catalog No. NC9203301
Encompass_Preferred
The MVE Biological Solutions Roller Base is a durable 5-wheel dolly that enhances mobility for cryogenic dewars and freezers, facilitating the safe transport of heavy units in laboratory environments. With a 20.0 in internal diameter, it is designed specifically for compatibility with various MVE cryogenic models, setting it apart through its sturdy construction and superior functionality.
Catalog No. NC9203301 Supplier MVE BIOLOGICAL SOLUTIONS Supplier No. 10937411
The MVE Biological Solutions Roller Base is a durable 5-wheel dolly that enhances mobility for cryogenic dewars and freezers, facilitating the safe transport of heavy units in laboratory environments. With a 20.0 in internal diameter, it is designed specifically for compatibility with various MVE cryogenic models, setting it apart through its sturdy construction and superior functionality.