Learn More
Labrepco Inc ROLLER BASE FOR LS750 LABSYSTE
SDP
Catalog No. 509020681
Encompass_Preferred
Roller Base for LS750, HC34, HC35, VHC35, XT34, LD35, & LD50 Liquid Nitrogen Systems. The roller base is a sturdy, aluminum, 5-wheel dolly that enables movement of freezers and dewars easily and safely.
Catalog No. 50-902-0681 Supplier Labrepco Inc Supplier No. R0338C00
Roller Base for LS750, HC34, HC35, VHC35, XT34, LD35, & LD50 Liquid Nitrogen Systems. The roller base is a sturdy, aluminum, 5-wheel dolly that enables movement of freezers and dewars easily and safely.