Learn More
Bellco Glass, Inc. Roller Bottle,110X270mm
SDP
Catalog No. 501946074
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass.. MADE IN THE USA 38 mm deep-skirt, nontoxic, rubber lined phenolic screw cap Dimensions: 110x270mm Area (cm2): 670 Replacement caps: 2029 series
Catalog No. 50-194-6074 Supplier Bellco Glass, Inc. Supplier No. 773051270
Manufactured from Type 1 Class A Borosilicate Glass.. MADE IN THE USA 38 mm deep-skirt, nontoxic, rubber lined phenolic screw cap Dimensions: 110x270mm Area (cm2): 670 Replacement caps: 2029 series