Learn More
Bellco Glass, Inc. Roller Bottle, 110x285mm
SDP
Catalog No. 501946068
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass.. MADE IN THE USA 38 mm deep-skirt, nontoxic, rubber lined phenolic screw cap Replacement cap: 2029-0038a Area (cm2): 670 Replacement caps: 2029 series Dimensions: 110x285mm
Catalog No. 50-194-6068 Supplier Bellco Glass, Inc. Supplier No. 773038285
Manufactured from Type 1 Class A Borosilicate Glass.. MADE IN THE USA 38 mm deep-skirt, nontoxic, rubber lined phenolic screw cap Replacement cap: 2029-0038a Area (cm2): 670 Replacement caps: 2029 series Dimensions: 110x285mm