Learn More
Bellco Glass, Inc. Roller Bottle, 110x440mm
SDP
Catalog No. 501946069
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass.. MADE IN THE USA 38 mm deep-skirt, nontoxic, rubber lined phenolic screw cap Dimensions: 110x440mm Area (cm2): 670 Replacement caps: 2029 series
Catalog No. 50-194-6069 Supplier Bellco Glass, Inc. Supplier No. 773038440
Manufactured from Type 1 Class A Borosilicate Glass.. MADE IN THE USA 38 mm deep-skirt, nontoxic, rubber lined phenolic screw cap Dimensions: 110x440mm Area (cm2): 670 Replacement caps: 2029 series