Learn More
Bellco Glass, Inc. Roller Bottle, 110x570mm With GL45 Blue PP Cap
SDP
Catalog No. 501945995
Encompass_Preferred
Borosilicate glass, type 1, class a45 mm polypropylene screw cap provides secure sealPolypropylene pouring ring allows drip free pouring and protects rimReplacement cap: 5637-00045; pouring ring: 5637-10045Area (cm2): 1585, OD x OAL: 110 x 570mm. MADE IN THE USA
Catalog No. 50-194-5995 Supplier Bellco Glass, Inc. Supplier No. 771545585
Borosilicate glass, type 1, class a45 mm polypropylene screw cap provides secure sealPolypropylene pouring ring allows drip free pouring and protects rimReplacement cap: 5637-00045; pouring ring: 5637-10045Area (cm2): 1585, OD x OAL: 110 x 570mm. MADE IN THE USA