Learn More
Bellco Glass, Inc. Roller Bottle,120x 285mm With Indented Bottom
SDP
Catalog No. 501946073
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass.. MADE IN THE USA 38 mm deep-skirt, nontoxic, rubber lined phenolic screw cap Dimensions: 120x285mm Area (cm2): 835 Replacement caps: 2029 series
Catalog No. 50-194-6073 Supplier Bellco Glass, Inc. Supplier No. 773051120
Manufactured from Type 1 Class A Borosilicate Glass.. MADE IN THE USA 38 mm deep-skirt, nontoxic, rubber lined phenolic screw cap Dimensions: 120x285mm Area (cm2): 835 Replacement caps: 2029 series