Learn More
CELLTREAT Scientific Products 2000mL Non-Treated Roller Bottle, Non-Vented Cap, Sterile
SDP
Catalog No. 50550485
Encompass_Preferred
CELLTREAT Scientific Products Non-treated 2000mL Roller Bottle. Large knurled cap for easy grip. Manufactured from medical grade virgin polystyrene. Easy-to-read black printed graduations. Non-Treated Roller Bottles can be used for suspension cell culture and large capacity liquid storage. Vent caps available separately for use with open systems. Non-vented cap. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individual bag, 12/case.
Catalog No. 50-550-485 Supplier CELLTREAT Scientific Products Supplier No. 229584
CELLTREAT Scientific Products Non-treated 2000mL Roller Bottle. Large knurled cap for easy grip. Manufactured from medical grade virgin polystyrene. Easy-to-read black printed graduations. Non-Treated Roller Bottles can be used for suspension cell culture and large capacity liquid storage. Vent caps available separately for use with open systems. Non-vented cap. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individual bag, 12/case.