Learn More
CELLTREAT Scientific Products 490cm Tissue Culture Treated Roller Bottle, Vented Cap, Sterile
SDP
Catalog No. 50201990
Encompass_Preferred
CELLTREAT Scientific Products 490cm¿ Roller Bottle. Large knurled cap for easy grip. Vented cap for open systems . Vent caps feature 0.22¿m hydrophobic membrane. Manufactured from medical grade virgin polystyrene. Tissue Culture Treated Roller Bottles have uniform hydrophilic surface ideal for consistent cell attachment. Easy-to-read black printed graduations. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile in individual bags, 24/case.
Catalog No. 50-201-990 Supplier CELLTREAT Scientific Products Supplier No. 229383
CELLTREAT Scientific Products 490cm¿ Roller Bottle. Large knurled cap for easy grip. Vented cap for open systems . Vent caps feature 0.22¿m hydrophobic membrane. Manufactured from medical grade virgin polystyrene. Tissue Culture Treated Roller Bottles have uniform hydrophilic surface ideal for consistent cell attachment. Easy-to-read black printed graduations. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile in individual bags, 24/case.