Learn More
CELLTREAT Scientific Products 850cm Tissue Culture Treated Roller Bottle, Non-Vented Cap, Sterile
SDP
Catalog No. 50201991
Encompass_Preferred
CELLTREAT Scientific Products 850cm¿ Roller Bottle. Large knurled cap for easy grip. Manufactured from medical grade virgin polystyrene. Tissue Culture Treated Roller Bottles have uniform hydrophilic surface ideal for consistent cell attachment. Easy-to-read black printed graduations. Non-vented cap. RNase/Dnase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile in individual bags, 12/case.
Catalog No. 50-201-991 Supplier CELLTREAT Scientific Products Supplier No. 229384
CELLTREAT Scientific Products 850cm¿ Roller Bottle. Large knurled cap for easy grip. Manufactured from medical grade virgin polystyrene. Tissue Culture Treated Roller Bottles have uniform hydrophilic surface ideal for consistent cell attachment. Easy-to-read black printed graduations. Non-vented cap. RNase/Dnase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile in individual bags, 12/case.