Learn More
Bellco Glass, Inc. Roller Bottle Brush For 110mm OD bottles
SDP
Catalog No. 501946077
Encompass_Preferred
Roller bottle brush for 110 mm od roller bottles w/51 mm opening.. MADE IN THE USANylon bristles for easy cleaning of hard-to0-reach surfaces. Useable length: 690 MMNOTE: to keep nylon bristles straight & effective dip flattened brush in hot water - then shake outMATERIAL:•Plastic handle •Plastic tube covering shaft •#8 wire - nylon
Catalog No. 50-194-6077 Supplier Bellco Glass, Inc. Supplier No. 773111111
Roller bottle brush for 110 mm od roller bottles w/51 mm opening.. MADE IN THE USANylon bristles for easy cleaning of hard-to0-reach surfaces. Useable length: 690 MMNOTE: to keep nylon bristles straight & effective dip flattened brush in hot water - then shake outMATERIAL:•Plastic handle •Plastic tube covering shaft •#8 wire - nylon