Learn More
CELLTREAT Scientific Products CAP ONLY, Vented Roller Bottle Cap, Sterile
SDP
Catalog No. 50550486
Encompass_Preferred
CELLTREAT Scientific Products Roller Bottle, Cap Only. Large knurled cap for easy grip. Vented cap for open systems. Vent caps feature 0.22¿m hydrophobic membrane. RNase/Dnase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 1/bag, 24/case.
Catalog No. 50-550-486 Supplier CELLTREAT Scientific Products Supplier No. 229391
CELLTREAT Scientific Products Roller Bottle, Cap Only. Large knurled cap for easy grip. Vented cap for open systems. Vent caps feature 0.22¿m hydrophobic membrane. RNase/Dnase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 1/bag, 24/case.