Learn More
CELLTREAT Scientific Products 1900cm Expanded Surface Tissue Culture Treated Roller Bottle, Vented Cap, Sterile
SDP
Catalog No. 50550483
Encompass_Preferred
CELLTREAT Scientific Products 1900cm¿ Expanded Surface Roller Bottle. Expanded Surface Roller Bottle increases surface area for cell culture. Large knurled cap for easy grip. Vented cap for open systems. Vent caps feature 0.22¿m hydrophobic membrane. Manufactured from medical grade virgin polystyrene. Tissue Culture Treated Roller Bottles have uniform hydrophilic surface ideal for consistent cell attachment. Easy-to-read black printed graduations. RNase/DNase free and non-pyrogenic. Every case is printed with the lot numer for quality traceability. Packaged sterile in individual bags, 12/case.
Catalog No. 50-550-483 Supplier CELLTREAT Scientific Products Supplier No. 229387
CELLTREAT Scientific Products 1900cm¿ Expanded Surface Roller Bottle. Expanded Surface Roller Bottle increases surface area for cell culture. Large knurled cap for easy grip. Vented cap for open systems. Vent caps feature 0.22¿m hydrophobic membrane. Manufactured from medical grade virgin polystyrene. Tissue Culture Treated Roller Bottles have uniform hydrophilic surface ideal for consistent cell attachment. Easy-to-read black printed graduations. RNase/DNase free and non-pyrogenic. Every case is printed with the lot numer for quality traceability. Packaged sterile in individual bags, 12/case.