Learn More
Microsolv Technology Corporation Roller for sealing Film or Mats in the U-2D Micro-Sample Management System. 1 EA.
Catalog No. 502270501
Encompass_Preferred
Complete application of Sealing Films or Mats by rolling over the Film or Mats with this Rolling Pin. Assure that a secure uniform seal around all wells and inserts are complete by an easy burnishing action that requires a small amount of pressure. Use with the U-2D Brand Micro-Sample Management System or with other 96 Well Plates.
This item is not returnable. View return policy
Catalog No. 50-227-0501 Supplier Microsolv Technology Corporation Supplier No. 95296ROLLER
Complete application of Sealing Films or Mats by rolling over the Film or Mats with this Rolling Pin. Assure that a secure uniform seal around all wells and inserts are complete by an easy burnishing action that requires a small amount of pressure. Use with the U-2D Brand Micro-Sample Management System or with other 96 Well Plates.