Learn More
Grainger Roller Guide Bottom 3-way 17 5/8 in
Catalog No. 502765681
Encompass_Preferred
The Grainger 16X580 is a 16-inch x 5/8-inch open end wrench made of chrome vanadium steel with a black oxide finish. It has a 15 degree offset head and meets ASME B107.300-2010 standards. This wrench is designed for turning hex nuts and bolts in tight spaces. The slim head profile allows access where other wrenches can't reach. Overall length is 22 inches and it weighs approximately 4 pounds. This is a durable, heavy-duty wrench suitable for industrial applications.
Catalog No. 50-276-5681 Supplier Grainger Supplier No. 16X580
The Grainger 16X580 is a 16-inch x 5/8-inch open end wrench made of chrome vanadium steel with a black oxide finish. It has a 15 degree offset head and meets ASME B107.300-2010 standards. This wrench is designed for turning hex nuts and bolts in tight spaces. The slim head profile allows access where other wrenches can't reach. Overall length is 22 inches and it weighs approximately 4 pounds. This is a durable, heavy-duty wrench suitable for industrial applications.